Phosphatidylbutanol ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
:
Formula:C₄₁H₇₈O₈P
:731.1 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together