BML-288
25 mg
Catalog # ENZOBMLPD2000025
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
PDE2 inhibitor
Potent (IC50=40nM) and selective (290X selectivity over other PDE isoforms) inhibitor of phosphodiesterase Type II (PDE2). Displayed no significant level of binding to neither a collection of 54 receptors and ion channels, nor to a panel of 30 kinases. BML-288 demonstrated no inhibition of 5-lipoxygenase (5-LO) or cyclooxygenase (COX-1). BML-288 is more potent and selective than EHNA (BML-PD129).
- CAS Number:851681-89-5
- Formula:C₁₄H₁₀N₄O₃S
- Molecular Weight:314.32 g/mol
Specifications
Specifications
- Pk:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



