Luffariellolide (from Luffariella sp.) ≥98% (by TLC)
Catalog # ENZOBMLST3100001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:386.6 g/mol
Storage Temperature:Freezer
CAS Number:111149-87-2
Formula:C₂₅H₃₈O₃