Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Apstatin
Apstatin ≥95% (by HPLC)
Apstatin ≥95% (by HPLC)
Apstatin ≥95% (by HPLC)
Catalog # ENZOBMLPI1450001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Apstatin ≥95% (by HPLC)
Catalog # ENZOBMLPI1450001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
459.5 g/mol
Storage Temperature:
Freezer
CAS Number:
160470-73-5
Formula:
C₂₃H₃₃N₅O₅
Product Details