Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SecinH3
SecinH3 ≥99% (by HPLC)
SecinH3 ≥99% (by HPLC)
ENZOENZCHM1140005
:
ENZO LIFE SCIENCES
:
SecinH3 ≥99% (by HPLC)
ENZOENZCHM1140005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
:
:
460.5 g/mol
:
853625-60-2
:
C₂₄H₂₀N₄O₄S
:
MFCD10687099