Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SANT-1
SANT-1 ≥99% (by HPLC)
SANT-1 ≥99% (by HPLC)
SANT-1 ≥99% (by HPLC)
Catalog # ENZOENZCHM1200001
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
SANT-1 ≥99% (by HPLC)
Catalog # ENZOENZCHM1200001
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
373.5 g/mol
CAS Number:
304909-07-7
Formula:
C₂₃H₂₇N₅
MDL Number:
MFCD01827306
Product Details