Order Entry
Netherlands
ContactUsLinkComponent
MMP-​9 inhibitor I ≥97% (by HPLC)
MMP-​9 inhibitor I ≥97% (by HPLC)
Catalog # ENZOENZCHM1330001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
MMP-​9 inhibitor I ≥97% (by HPLC)
Catalog # ENZOENZCHM1330001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :511.6 g/mol
CAS Number:1177749-58-4
MDL Number:MFCD22581508