Order Entry
Netherlands
ContactUsLinkComponent
Ikarugamycin ≥95% (by HPLC, TLC)
Ikarugamycin ≥95% (by HPLC, TLC)
Catalog # ENZOBMLEI3130500
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Ikarugamycin ≥95% (by HPLC, TLC)
Catalog # ENZOBMLEI3130500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :478.6 g/mol
Storage Temperature:Freezer
CAS Number:36531-78-9
Formula:C₂₉H₃₈N₂O₄