Order Entry
Netherlands
ContactUsLinkComponent
Chromomycin A3 (from Streptomyces griseus) ≥98% (by TLC)
Chromomycin A3 (from Streptomyces griseus) ≥98% (by TLC)
Catalog # ENZOBMLGR3020010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Chromomycin A3 (from Streptomyces griseus) ≥98% (by TLC)
Catalog # ENZOBMLGR3020010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :1183.30 g/mol
Storage Temperature:Freezer
CAS Number:7059-24-7
Formula:C₅₇H₈₂O₂₆
MDL Number:MFCD00043151