Order Entry
Netherlands
ContactUsLinkComponent
 
Toxic
25-Hydroxyvitamin D₂ ≥98% (by HPLC)
Catalog # ENZOBMLDM1010001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Toxic
25-Hydroxyvitamin D₂ ≥98% (by HPLC)
Catalog # ENZOBMLDM1010001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :412.65 g/mol
UN:2811
ADR:6.1,II
 :GHS06
CAS Number:21343-40-8
Formula:C₂₈H₄₄O₂
MDL Number:MFCD02683431