BML-111 ≥99%
Supplier: Enzo Life Sciences
About this item
Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).
Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).
Synonyms: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
- CAS Number:78606-80-1
- Formula:C₈H₁₆O₅
- Molecular Weight:192.2 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).
Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).
Synonyms: 5(S),6(R),7-Trihydroxyheptanoic acid methyl ester
- CAS Number:78606-80-1
- Formula:C₈H₁₆O₅
- Molecular Weight:192.2 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Purity:≥99%



