Order Entry
Netherlands
ContactUsLinkComponent
Farnesyl thiotriazole ≥98% (by TLC)
Farnesyl thiotriazole ≥98% (by TLC)
Catalogus # ENZOBMLPE1900010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Farnesyl thiotriazole ≥98% (by TLC)
Catalogus # ENZOBMLPE1900010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :305.5 g/mol
Storage Temperature:Freezer
CAS Number:156604-45-4
Formula:C₁₇H₂₆N₃S
MDL Number:MFCD00270863