Order Entry
Netherlands
ContactUsLinkComponent
4-Hydroxytriazolam ≥97% (by HPLC)
Irritant
Health
4-Hydroxytriazolam ≥97% (by HPLC)
Catalog # ENZOBMLJ1310005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Health
4-Hydroxytriazolam ≥97% (by HPLC)
Catalog # ENZOBMLJ1310005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥97% (HPLC)
  • Source:
    Synthetic

Specification Test Results

About this item

 :
 :359.2 g/mol
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:65686-11-5
Formula:C₁₇H₁₂Cl₂N₄O