Order Entry
Netherlands
ContactUsLinkComponent
Oligomycin A ≥97% (by HPLC, TLC)
Oligomycin A ≥97% (by HPLC, TLC)
Catalog # ENZOBMLCM1110005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Oligomycin A ≥97% (by HPLC, TLC)
Catalog # ENZOBMLCM1110005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :791.07 g/mol
Storage Temperature:Freezer
CAS Number:579-13-5
Formula:C₄₅H₇₄O₁₁
MDL Number:MFCD00065705