
3-Tropanyl-3,5-dichlorobenzoate ≥99% (by HPLC), white solid
Catalog # ENZOBMLNS5300050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- VPE:50 mg
- Seq:2370
Specifications
Specification Test Results
- Purity:≥99% (HPLC)
Specification Test Results
About this item
:
:314.206 g/mol
:GHS06
Storage Temperature:Kamertemperatuur
CAS Number:40796-97-2
Formula:C₁₅H₁₇Cl₂NO₂
MDL Number:MFCD00865533