2-Chloro-5-nitro-N-(4-pyridyl)benzamide ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:313516-66-4
:
Formula:C₁₂H₈ClN₃O₃
MDL Number:MFCD00121849
:277.7 g/mol
Storage Temperature:Ambient
Frequently Bought Together