Order Entry
Netherlands
ContactUsLinkComponent
6-Formylindolo[3,2-b]carbazole ≥95% (by HPLC), yellow solid
Irritant
6-Formylindolo[3,2-b]carbazole ≥95% (by HPLC), yellow solid
Catalog # ENZOBMLGR2060100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
6-Formylindolo[3,2-b]carbazole ≥95% (by HPLC), yellow solid
Catalog # ENZOBMLGR2060100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

Specification Test Results

  • Purity:
    ≥95% (HPLC)
  • Appearance:
    Yellow solid

Specification Test Results

About this item

 :
Melting Pt:>300 °C
 :284.32 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:172922-91-7
 :MFCD09879259
Formula:C₁₉H₁₂N₂O