Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Veratridine
Veratridine ≥98% (by TLC)
Veratridine ≥98% (by TLC)
ENZOBMLNA1250010
:
ENZO LIFE SCIENCES
New Product
:
Veratridine ≥98% (by TLC)
ENZOBMLNA1250010
:
ENZO LIFE SCIENCES
New Product
:
Pk:
10 mg
Seq:
2275
Environmentally Preferable:
Purity:
≥98% (TLC)
Appearance:
White to yellow solid.
:
:
673.8 g/mol
:
GHS06
:
GHS08
:
Freezer
:
71-62-5
:
C₃₆H₅₁NO₁₁