Order Entry
Netherlands
ContactUsLinkComponent
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
Catalog # ENZOBMLL1200005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
Catalog # ENZOBMLL1200005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :538.707 g/mol
Storage Temperature:Freezer
CAS Number:91575-58-5
Formula:C₂₆H₅₅N₂O₇P
MDL Number:MFCD00065891