Order Entry
Netherlands
ContactUsLinkComponent
LY 171883 ≥90% (by HPLC)
Irritant
LY 171883 ≥90% (by HPLC)
Catalog # ENZOBMLRA1010010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
LY 171883 ≥90% (by HPLC)
Catalog # ENZOBMLRA1010010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥90% (HPLC)

Specification Test Results

About this item

 :
 :318.377 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:88107-10-2
Formula:C₁₆H₂₂N₄O₃
MDL Number:MFCD00211057