Order Entry
Netherlands
ContactUsLinkComponent
JC-1 (iodide) ≥95% (by HPLC), Ultrapure
JC-1 (iodide) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52304
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
JC-1 (iodide) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52304
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :652.223 g/mol
Storage Temperature:Freezer
CAS Number:3520-43-2
 :47729-63-5
Formula:C₂₅H₂₇Cl₄IN₄
MDL Number:MFCD00188475