Order Entry
Netherlands
ContactUsLinkComponent
Z-Asp-2,6-dichlorobenzoyloxymethylketone ≥98% (by TLC)
Z-Asp-2,6-dichlorobenzoyloxymethylketone ≥98% (by TLC)
Catalog # ENZOALX260029M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Z-Asp-2,6-dichlorobenzoyloxymethylketone ≥98% (by TLC)
Catalog # ENZOALX260029M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :454.256 g/mol
CAS Number:153088-73-4
Formula:C₂₀H₁₇Cl₂NO₇
MDL Number:MFCD00674885