Gallocyanine ≥98% (by TLC)
Catalog # ENZOBMLWN1180500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:336.73 g/mol
CAS Number:1562-85-2
Formula:C₁₅H₁₃ClN₂O₅
MDL Number:MFCD00011926