

Docetaxel ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:114977-28-5
:
Formula:C₄₃H₅₃NO₁₄
MDL Number:MFCD00800737
:807.89 g/mol
:GHS07
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (HPLC)
Specification Test Results
Frequently Bought Together