Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Minoxidil
Minoxidil ≥98% (by HPLC)
Minoxidil ≥98% (by HPLC)
ENZOBMLKC1250500
:
Enzo Life Sciences
:
Minoxidil ≥98% (by HPLC)
ENZOBMLKC1250500
:
Enzo Life Sciences
:
Pk:
500 mg
Seq:
2725
Purity:
≥98% (HPLC)
:
:
209.253 g/mol
:
14,06203
:
GHS07
:
38304-91-5
:
C₉H₁₅N₅O
:
MFCD00063409