Order Entry
Netherlands
ContactUsLinkComponent
N-Acetyl-L-cysteine
N-Acetyl-L-cysteine
Catalog # ENZOALX105005G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
N-Acetyl-L-cysteine
Catalog # ENZOALX105005G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :163.2 g/mol
Merck Index:13,00090
Boiling Pt:371 °C (1013 hPa)
Melting Pt:106…108 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-498-3
CAS Number:616-91-1
Formula:C₅H₉NO₃S
MDL Number:MFCD00004880