Pellicon® Capsule with Ultracel® Membrane
MILXPCC300C01
: Merck
- Filtration area:0,1 m² cm²
- Type:Ultrafiltration
- Description:Pellicon® capsule with Ultracel® membrane, 300 kDa, C screen
- Inlet/outlet connection:Feed: ³/₄" sanitary flange
Retentate: ³/₄" sanitary flange
Permeate: ³/₄" sanitary flange - Ø×L:380×353 mm
- Max. operating temperature:Inlet pressure: 80 psi (5,5 bar) at 4...30 °C
- Max. operating pressure:Forward transmembrane pressure (TMP): 45 psi (3,1 bar) at 4...30 °C
- Pk:1
Applications: Viral gene therapies, vaccines, concentration, diafiltration, buffer exchange, mRNA.
- Sterilised by gamma irradiation
Flow rate range feed: 4 to 6 L/min per m²
pH range: 2 to 13
Hold-up volume: 28 ml in feed channel, 60 ml in permeate channel
Chemistry: Composite regenerated cellulose (CRC)
Device material: Housing/Core - PPO/PS blend
Potting: BPA-free epoxy, polyurethane
Screen material: Polypropylene (PP), polyester
Seal material: EPDM, thermoplastic elastomer
Storage temperature: 15 to 30 °C
TOC: Meets TOC ≤5 ppm in retentate effluent, after a WFI flush of 20 L/m²