Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
PP.
- High chemical and temperature resistance
- Available for Eppendorf systems (except 211-2522: Vitatron and 211-2524: Roche)
- Free of detectable DNase, RNase, human DNA, non-pyrogenic
PP.
- High chemical and temperature resistance
- Available for Eppendorf systems (except 211-2522: Vitatron and 211-2524: Roche)
- Free of detectable DNase, RNase, human DNA, non-pyrogenic
- Catalog No:
- 211-2522
- 211-2520
- 211-2521
- 525-0489
- 525-0490
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Product Family Options
With or without caps
- CapacityColourTypeVersionRCF max.Packed
- 0,5 mlColourlesswith attached lidgraduated, conical bottom, non sterile51 400×g1000/bag, 10 000/box1 * 10.000 ST609,00 €
Specifications
Cat. No.211-2522Environmentally PreferableEnvironmental certification - 1,5 mlColourlesswith attached lidgraduated, conical bottom, non sterile18 000×g500/bag, 4000/box1 * 4.000 ST59,70 €
Low Manufacturing Impact Specifications
Cat. No.211-2520Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 2,2 mlColourlesswith attached lidgraduated, conical bottom, non sterile16 000×g500/bag, 4000/box1 * 4.000 ST228,00 €
Low Manufacturing Impact Specifications
Cat. No.211-2521Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact
With thread, without screw caps
- CapacityColourTypeVersionRCF max.Packed
- 1,5 mlColourlesswith thread, without screw capgraduated, conical bottom, non sterile20 000×g500/bag, 5000/box1 * 500 ST37,50 €
Specifications
Cat. No.525-0489Environmentally PreferableEnvironmental certification - 1,5 mlColourlesswith thread, without screw capgraduated, skirted, non sterile20 000×g500/bag, 5000/box1 * 500 ST39,40 €
Specifications
Cat. No.525-0490Environmentally PreferableEnvironmental certification
Screw caps for microcentrifuge tubes
- ColourDescriptionFor tubes