Order Entry
Netherlands
ContactUsLinkComponent
Barium hydroxide 0.05 mol/l (0.1 N) in aqueous solution
Barium hydroxide 0.05 mol/l (0.1 N) in aqueous solution
Catalog # ROTHN054.1
Supplier:  Roth Carl
CAS Number:  
Barium hydroxide 0.05 mol/l (0.1 N) in aqueous solution
Catalog # ROTHN054.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :171.34 g/mol
EINECS:241-234-5
CAS Number:17194-00-2
Formula:H₂BaO₂
MDL Number:MFCD00003443