2-Hydroxyisobutyric acid 98%
:594-61-6
:
:C₄H₈O₃
:MFCD00004459
:104.11 g/mol
:84 °C (1.5 mmHg)
:77…81 °C
:209-848-8
:GHS07
:GHS05
:Ambient
- Appearance:White to cream Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:75 to 81 °C
- Titration with NaOH:≥97.5 to 102.5 %
Frequently Bought Together