

2-Fluoro-4-hydroxybenzonitrile ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:82380-18-5
:
Formula:C₇H₄FNO
MDL Number:MFCD00192177
:137.11 g/mol
Boiling Pt:261 °C (1013 hPa)
Melting Pt:123…126 °C
Density:1.450
Storage Temperature:Ambient
EINECS:422-810-7
ADR:9,III
:GHS05
:GHS07
Related Information
Frequently Bought Together