0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Sepharose™ CL-4B
Sepharose™ CL-4B, cross-linked, Supelco®
Sepharose™ CL-4B, cross-linked, Supelco®
Catalog # SUPLCL4B200-100ML
Supplier:
Merck
CAS Number:
Expand image
Sepharose™ CL-4B, cross-linked, Supelco®
Catalog # SUPLCL4B200-100ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Productdetails
:
CAS Number:
61970-08-9
MDL Number:
MFCD00148279
Productdetails