Atto 594 maleimide ≥90% (by HPLC) (Coupling efficiency with N-Acetylcysteine), BioReagent suitable for fluorescence, Supelco®
Catalog # SUPL08717-1MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:928.09 g/mol
MDL Number:MFCD07370575