Order Entry
Netherlands
ContactUsLinkComponent
Atto 633 NHS ester ≥90% (by HPLC), BioReagent suitable for fluorescence, Supelco®
Atto 633 NHS ester ≥90% (by HPLC), BioReagent suitable for fluorescence, Supelco®
Catalog # SUPL01464-1MG-F
Supplier:  Merck
CAS Number:  
Atto 633 NHS ester ≥90% (by HPLC), BioReagent suitable for fluorescence, Supelco®
Catalog # SUPL01464-1MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
MDL Number:MFCD07784939