Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD72.1 Mouse Monoclonal Antibody [clone:10.1.D2]
Anti-CD72.1 Mouse Monoclonal Antibody [clone:10.1.D2]
Catalog # PRSI98-840
Supplier:  ProSci Inc.
Anti-CD72.1 Mouse Monoclonal Antibody [clone:10.1.D2]
Catalog # PRSI98-840
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD72.1
  • Clonality:
    Monoclonal
  • Clone:
    10.1.D2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Storage buffer:
    Supplied in borate buffered saline, pH 8,2
  • Storage temperature:
    Store vial at 2...8 °C
  • Pk:
    0,5 mg

Specifications

About this item

Type: Primary
Antigen: CD72.1
Clonality: Monoclonal
Clone: 10.1.D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse