Order Entry
Netherlands
ContactUsLinkComponent
Anti-M (SARS-CoV-2 (COVID-19)) Rabbit Polyclonal Antibody
Anti-M (SARS-CoV-2 (COVID-19)) Rabbit Polyclonal Antibody
Catalog # PRSI10-516
Supplier:  ProSci Inc.
Anti-M (SARS-CoV-2 (COVID-19)) Rabbit Polyclonal Antibody
Catalog # PRSI10-516
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    SARS-CoV-2 (COVID-19)
  • Antigen symbol:
    M
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Format:
    Liquid
  • Gene ID:
    43740571
  • Antigen synonyms:
    Membrane protein|E1 Glycoprotein|SARS-CoV-2 (COVID-19, 2019-nCoV)|M protein
  • Modification:
    None
  • Accession no.:
    QHD43419.1
  • Storage buffer:
    PBS with 0,02% sodium azide, 10% glycerol, pH 7,2
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Concentration:
    Batch dependent
  • Immunogen:
    Synthetic peptide located within the center of SARS-CoV-2 (COVID-19) Membrane
  • Purification:
    Affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE)
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

SARS-CoV-2 (COVID-19) membrane antibody.

Optimal dilutions for each application to be determined by the researcher.

Type:
Antigen: M (SARS-CoV-2 (COVID-19))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus