Pesticide reference standard, 2,5-Dichlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12505800ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:19398-61-9
- Description:2,5-Dichlorotoluene 100 µg/ml in methanol
- Formula:C₇H₆Cl₂
- Description:2,5-Dichlorotoluene