Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, 2,6-Dichlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 2,6-Dichlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12505900ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,6-Dichlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12505900ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    118-69-4
  • Description:
    2,6-Dichlorotoluene 100 µg/ml in methanol
  • Formula:
    C₇H₆Cl₂
  • Description:
    2,6-Dichlorotoluene

Specifications

About this item