Anti-GPRC5A/RAI3 Mouse Monoclonal Antibody [clone: 481906] (Alexa Fluor 405)
NOVUFAB5239V
- Antibody type:Primary
- Antigen symbol:GPRC5A/RAI3
- Clonality:Monoclonal
- Clone:481906
- Conjugation:Alexa Fluor® 405
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen synonyms:RAIG1RAIG-1|GPCR5A|retinoic acid responsive|Retinoic acid-induced gene 1 protein|Orphan G-protein-coupling receptor PEIG-1|RAIG1|retinoic acid induced 3|G protein-coupled receptor, family C, group 5, member A|GPRC5A|RAI3|RAI3G-protein coupled receptor family C group 5 member A|retinoic acid-induced protein 3
- Storage buffer:Supplied 0,2 mg/ml in a saline solution containing BSA and Sodium Azide.
- Molecular weight:35 kDa
- Storage temperature:Store the unopened product at 2...8 °C.
- Immunogen:NS0 mouse myeloma cell line transfected with human GPRC5A
Met1-Ser357
Accession # Q8NFJ5 - Size:100 µg
- Pk:0,1 mL
G-protein coupled receptor family C group 5 member A (GPRC5A), also known as Retinoic acid-induced gene 1 protein (RAIG-1), is a member of the G-protein coupled receptor 3 family.
- Has a molecular weight of approximately 35 kDa and contains seven transmembrane domains
Human GPRC5A shares 76% and 74% aa sequence identity with mouse and rat GPRC5A, respectively. It is expressed at high levels in fetal and adult lung and is also present in fetal kidney and adult placenta, kidney, prostate, testis, ovary, small intestine, colon, stomach, and spinal chord at lower levels.
Primary antibodies are guaranteed for 1 year from date of receipt.
: For research use only and is not approved for use in humans or in clinical diagnosis.
Type: Primary
Antigen: GPRC5A/RAI3
Clonality: Monoclonal
Clone: 481906
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human