Anti-14-3-3 Binding Motif Phosphate Rabbit Polyclonal Antibody
Catalog # CSTA9601S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 binding motif phosphate
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Multiple
- Western blot:Yes
- Sequence:XXXXRSXsXPXXXXC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: 14-3-3 binding motif phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Any species