Anti-Rb (Ser807/Ser811) Phosphate Rabbit Monoclonal Antibody [clone: D20B12] (Alexa Fluor 555)
Catalog # CSTA8957S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Rb (Ser807/Ser811) phosphate
- Clonality:Monoclonal
- Clone:D20B12
- Conjugation:Alexa Fluor® 555
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Sequence:GNIYIsPLKsPYKISC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Rb (Ser807/Ser811) phosphate
Clonality: Monoclonal
Clone: D20B12
Conjugation: Alexa Fluor 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey