Anti-Akt-Phosphate Substrate (Ser/Thr) Phosphate Rabbit Monoclonal Antibody [clone: 110B7E] (HRP)
CSTA6950S
- Antibody type:Primary
- Antigen symbol:Akt-phosphate Substrate (Ser/Thr) phosphate
- Clonality:Monoclonal
- Clone:110B7E
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Sequence:CXXXRXRXXtXXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: Akt-phosphate Substrate (Ser/Thr) phosphate
Clonality: Monoclonal
Clone: 110B7E
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: