Anti-HER3/ErbB3 (Tyr1197) Phosphate Rabbit Monoclonal Antibody [clone: C56E4]
Catalog # CSTA4561S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HER3/ErbB3 (Tyr1197) phosphate
- Clonality:Monoclonal
- Clone:C56E4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequence:CEEDEDEEyE
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HER3/ErbB3 (Tyr1197) phosphate
Clonality: Monoclonal
Clone: C56E4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse