Anti-A-Raf Rabbit Polyclonal Antibody
Catalog # CSTA4432S
Marketsource
Supplier: Cell Signaling
Jump to:
Product Details & Documents
Type: Primary
Antigen: A-Raf
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Specifications
- Seq:02
- Western blot:Yes
- Cat. No.:CSTA4432S
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Storage temperature:−20 °C
- Host:Rabbit
- Sequence:CGRKSPHSK(S/L)P(A/S)EQRERKSL
- Antigen symbol:A-Raf
- Shipping temperature:Ambient
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



