Anti-BCL-10 Rabbit Monoclonal Antibody [clone: C78F1]
Catalog # CSTA4237S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:BCL-10
- Clonality:Monoclonal
- Clone:C78F1
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CRTSSRKRAGKLLDYLQENPKGLDTL
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BCL-10
Clonality: Monoclonal
Clone: C78F1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat