Anti-Synuclein-alpha Rabbit Monoclonal Antibody [clone: D37A6]
Catalog # CSTA4179S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Synuclein-alpha
- Clonality:Monoclonal
- Clone:D37A6
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CKDQMGKGEEGYPQEGILED
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Synuclein-alpha
Clonality: Monoclonal
Clone: D37A6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat