Anti-CSF1R (Tyr723) Phosphate Rabbit Monoclonal Antibody [clone: 49C10]
CSTA3155S
- Antibody type:Primary
- Antigen symbol:CSF1R (Tyr723) phosphate
- Clonality:Monoclonal
- Clone:49C10
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequence:CSQGVDTyVEMRPV
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: CSF1R (Tyr723) phosphate
Clonality: Monoclonal
Clone: 49C10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse