Anti-ERK1 (Thr202/Tyr204) Phosphate Rabbit Monoclonal Antibody [clone: 197G2] (Alexa Fluor 488)
Catalog # CSTA13214S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ERK1 (Thr202/Tyr204) phosphate
- Clonality:Monoclonal
- Clone:197G2
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Pig,Monkey,Human,Mink,D. melanogasterm,Rat,Mouse
- Antigen synonyms:ERK2 (Thr185/Tyr187) phosphate
- Sequence:TGFLtEyVATRWYRC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ERK1 (Thr202/Tyr204) phosphate
Clonality: Monoclonal
Clone: 197G2
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Mink, D. melanogasterm, Zebrafish, Pig