Anti-H3 (Lys4) dimethylate Rabbit Monoclonal Antibody [clone: C64G9]
Catalog # CSTA9725S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:H3 (Lys4) dimethylate
- Clonality:Monoclonal
- Clone:C64G9
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- ChIP:Yes
- Sequence:ART(K-di-methy)QTARKSGGC
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Tested applications:CUT&RUN
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: H3 (Lys4) dimethylate
Clonality: Monoclonal
Clone: C64G9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey