Anti-Casp3 (Asp175) cleaved Rabbit Monoclonal Antibody [clone: D3E9] (Alexa Fluor 488)
Catalog # CSTA9603S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Casp3 (Asp175) cleaved
- Clonality:Monoclonal
- Clone:D3E9
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Storage temperature:+4 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Casp3 (Asp175) cleaved
Clonality: Monoclonal
Clone: D3E9
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse